Assay Details
| Gene Symbol: Fam46c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142952.1 | NP_001136424.1 | 1 - 2 | 102 | 71 | |
| GenBank mRNA | BC037072.1 | - | 1 - 2 | 92 | 71 | |
| BC046309.1 | - | 1 - 2 | 96 | 71 | ||
| BC120717.1 | - | 1 - 2 | 93 | 40129640 | 71 | |
| BC120719.1 | - | 1 - 2 | 93 | 40129642 | 71 | |
| BY170905.1 | - | 1 - 2 | 102 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120717.1, BC120719.1, NM_001142952.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | BC037072.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | BC046309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | NM_001142952.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | AK015259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | BC037072.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | BC046309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | NM_001142952.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P447976 | AK015259.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106674 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134694 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111335 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | BC037072.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | BC046309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | NM_001142952.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | AK015259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | BC037072.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | BC046309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | NM_001142952.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P447976 | AK015259.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111335 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134694 | - | - |
| Whole Mouse Genome G4122A | A_51_P447976 | BC037072.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447976 | BC046309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447976 | NM_001142952.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447976 | AK015259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447976 | BC037072.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447976 | BC046309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447976 | NM_001142952.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447976 | AK015259.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111335 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02626820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02698297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03071113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02841031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03055702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02626822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02328594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02981208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02981215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03104417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03104422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03055701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02843586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03104424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02626815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02843593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22369482 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500612 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500613 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500611 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448021_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429682_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500610 | - | - |
| Mouse Genome 430 2.0 Array | 1429682_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429682_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74645_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74645_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300020138.mm.1 | NM_001142952.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020138.mm.1 | NM_001142952.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020138.mm.1 | TC0300002582.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020136.mm.1 | NM_001142952.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020136.mm.1 | NM_001142952.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020136.mm.1 | TC0300002582.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020137.mm.1 | NM_001142952.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020137.mm.1 | NM_001142952.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020137.mm.1 | TC0300002582.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429682_at | - | - |
| Mouse Expression Set 430 | 1429682_PM_at | - | - |
| Murine Genome U74v2 Set | 134703_PM_at | - | - |
| Murine Genome U74v2 Set | 134703_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429682_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429682_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 906955 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684205 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684205 | - | - |

Fam46c
Genomic Map