Assay Details
| Gene Symbol: Tspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009775.4 | NP_033905.3 | 2 - 3 | 278 | 75 | |
| GenBank mRNA | AA472650.1 | - | 2 - 3 | 277 | 75 | |
| AK149743.1 | - | 2 - 3 | 253 | 75 | ||
| AK150516.1 | - | 2 - 3 | 250 | 75 | ||
| AK151421.1 | - | 2 - 3 | 252 | 75 | ||
| AK151569.1 | - | 2 - 3 | 259 | 75 | ||
| AK151824.1 | - | 2 - 3 | 259 | 75 | ||
| AK152082.1 | - | 2 - 3 | 250 | 75 | ||
| AK152827.1 | - | 2 - 3 | 250 | 75 | ||
| AK152860.1 | - | 2 - 3 | 252 | 75 | ||
| AK153159.1 | - | 2 - 3 | 252 | 75 | ||
| AK153273.1 | - | 2 - 3 | 262 | 75 | ||
| AK156489.1 | - | 2 - 3 | 253 | 75 | ||
| AK165450.1 | - | 2 - 3 | 252 | 75 | ||
| AK165822.1 | - | 2 - 3 | 259 | 75 | ||
| AK166125.1 | - | 2 - 3 | 259 | 75 | ||
| AK172384.1 | - | 2 - 3 | 253 | 75 | ||
| BC002055.1 | - | 2 - 3 | 240 | 3493196 | 75 | |
| D21207.1 | - | 2 - 3 | 242 | 75 | ||
| L17306.1 | - | 1 - 2 | 207 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tspo
Genomic Map