Assay Details
| Gene Symbol: Zfp641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173769.3 | NP_776130.1 | 2 - 3 | 234 | 74 | |
| GenBank mRNA | AK036826.1 | - | 2 - 3 | 235 | 74 | |
| AK087530.1 | - | 2 - 3 | 282 | 74 | ||
| AK136402.1 | - | 2 - 3 | 312 | 74 | ||
| BC111896.1 | - | 2 - 3 | 234 | 40061302 | 74 | |
| BC112418.1 | - | 2 - 3 | 234 | 40061301 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp641
Genomic Map