| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300020880.mm.1 |
NM_001200028.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020880.mm.1 |
NM_001200028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020880.mm.1 |
NM_029604.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020880.mm.1 |
NM_029604.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020880.mm.1 |
TC0300002682.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020882.mm.1 |
NM_001200028.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020882.mm.1 |
NM_001200028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020882.mm.1 |
TC0300002682.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020879.mm.1 |
NM_001200028.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020879.mm.1 |
NM_001200028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020879.mm.1 |
NM_029604.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020879.mm.1 |
NM_029604.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020879.mm.1 |
TC0300002682.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020881.mm.1 |
NM_029604.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020881.mm.1 |
NM_029604.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020881.mm.1 |
TC0300002682.mm.1 |
MTA TC |