Assay Details
| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178919.4 | NP_849250.1 | 11 - 12 | 1812 | 84 | |
| GenBank mRNA | AK042427.1 | - | 11 - 12 | 1774 | 86 | |
| AK084606.1 | - | 11 - 12 | 1638 | 84 | ||
| AK085435.1 | - | 11 - 12 | 1765 | 84 | ||
| AK149916.1 | - | 11 - 12 | 1764 | 84 | ||
| AK149934.1 | - | 11 - 12 | 1813 | 84 | ||
| AK158371.1 | - | 10 - 11 | 1867 | 84 | ||
| AK171488.1 | - | 11 - 12 | 1793 | 84 | ||
| BC026448.1 | - | 10 - 11 | 1592 | 84 | ||
| BC038039.1 | - | 10 - 11 | 2314 | 84 | ||
| BC058973.1 | - | 7 - 8 | 2094 | 84 | ||
| BC079532.1 | - | 11 - 12 | 1708 | 6403184 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmf2
Genomic Map