Assay Details
| Gene Symbol: Mapk8ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021921.3 | NP_068740.3 | 2 - 3 | 487 | 107 | |
| GenBank mRNA | AF220195.1 | - | 2 - 3 | 206 | 107 | |
| AK014339.1 | - | 2 - 3 | 191 | 107 | ||
| AK158968.1 | - | 2 - 3 | 341 | 107 | ||
| BC029704.1 | - | 2 - 3 | 266 | 5359447 | 107 | |
| CA317486.1 | - | 2 - 3 | 462 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk8ip2
Genomic Map