Assay Details
| Gene Symbol: Panx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002005.2 | NP_001002005.2 | 2 - 3 | 2039 | 83 | |
| GenBank mRNA | DQ093579.1 | - | 2 - 3 | 1661 | 83 | |
| EU446266.1 | - | 2 - 3 | 2055 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422821 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422883 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058315 | DQ093579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058315 | NM_001002005.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978347 | DQ093579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978347 | NM_001002005.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346203 | AK054201.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346203 | XM_144122.7 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76392 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160924 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058315 | DQ093579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058315 | NM_001002005.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978347 | DQ093579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978347 | NM_001002005.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30422828 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160924 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08454336 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160924 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12204339 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12215157 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12204343 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422821 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30422884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30422814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12215154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12435421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12215145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12215148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12204340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12364264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12315682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12364259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11844675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12315670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12215150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12204338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12234982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12215157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12234955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11878900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30422821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11844676 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426156 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442204_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440350_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426150 | - | - |
| Mouse Genome 430 2.0 Array | 1440350_at | - | - |
| Mouse Genome 430 2.0 Array | 1442204_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440350_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442204_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500006727.mm.1 | NM_001002005.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006727.mm.1 | NM_001002005.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006727.mm.1 | TC1500000859.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500006729.mm.1 | NM_001002005.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006729.mm.1 | NM_001002005.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006729.mm.1 | TC1500000859.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500006728.mm.1 | NM_001002005.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006728.mm.1 | NM_001002005.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006728.mm.1 | TC1500000859.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500006731.mm.1 | NM_001002005.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006731.mm.1 | NM_001002005.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006731.mm.1 | TC1500000859.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500006725.mm.1 | NM_001002005.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006725.mm.1 | NM_001002005.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500006725.mm.1 | TC1500000859.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440350_PM_at | - | - |
| Mouse Expression Set 430 | 1442204_at | - | - |
| Mouse Expression Set 430 | 1442204_PM_at | - | - |
| Mouse Expression Set 430 | 1440350_at | - | - |
| Murine Genome U74v2 Set | 139495_at | - | - |
| Murine Genome U74v2 Set | 139495_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442204_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440350_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442204_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440350_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161563 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161563 | - | - |
| mouse-6 v1.1 (Illumina) | GI_50284540-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161563 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161563 | - | - |

Panx2
Genomic Map