Assay Details
| Gene Symbol: Dennd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027081.3 | NP_081357.2 | 1 - 2 | 250 | 74 | |
| GenBank mRNA | AK084555.1 | - | 1 - 2 | 243 | 74 | |
| AK154949.1 | - | 1 - 2 | 272 | 74 | ||
| BB658928.1 | - | 1 - 2 | 243 | 74 | ||
| BC139003.1 | - | 1 - 2 | 224 | 8862025 | 74 | |
| BY252285.1 | - | 1 - 2 | 252 | 74 | ||
| BY748566.1 | - | 1 - 2 | 267 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd6b
Genomic Map