Assay Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199198.2 | NP_954668.2 | 2 - 3 | 449 | 86 | |
| NR_028447.1 | 2 - 3 | 449 | 86 | |||
| NR_028448.1 | 2 - 3 | 449 | 86 | |||
| NR_028449.1 | 2 - 3 | 449 | 86 | |||
| GenBank mRNA | AK040235.1 | - | 2 - 3 | 401 | 86 | |
| AK045028.1 | - | 2 - 3 | 430 | 86 | ||
| AK154045.1 | - | 2 - 3 | 413 | 86 | ||
| BC013700.1 | - | 2 - 3 | 402 | 86 | ||
| BC064018.1 | - | 2 - 3 | 376 | 3980699 | 86 | |
| BU706947.1 | - | 2 - 3 | 190 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdac10
Genomic Map