Assay Details
| Gene Symbol: Tubgcp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163319.1 | NP_001156791.1 | 20 - 21 | 4809 | 82 | |
| GenBank mRNA | BC057626.1 | - | 5 - 6 | 1716 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp6
Genomic Map