Assay Details
| Gene Symbol: Tubgcp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163319.1 | NP_001156791.1 | 9 - 10 | 2166 | 67 | |
| GenBank mRNA | AK052441.1 | - | 9 - 10 | 2104 | 67 | |
| CF536834.1 | - | 3 - 4 | 319 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp6
Genomic Map