Assay Details
| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021423.3 | NP_067398.2 | 15 - 16 | 1891 | 68 | |
| GenBank mRNA | AB231013.1 | - | 15 - 16 | 1891 | 68 | |
| AB841411.1 | - | 5 - 6 | 855 | 68 | ||
| AB841412.1 | - | 5 - 6 | 855 | 68 | ||
| AJ245904.1 | - | 13 - 14 | 1745 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shank3
Genomic Map