Assay Details
| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021423.3 | NP_067398.2 | 5 - 6 | 600 | 92 | |
| GenBank mRNA | AB231013.1 | - | 5 - 6 | 92 | ||
| AJ245904.1 | - | 3 - 4 | 454 | 92 | ||
| KF612592.1 | - | 1 - 2 | 116 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shank3
Genomic Map