Assay Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011161.5 | NP_035291.4 | 5 - 6 | 537 | 75 | |
| GenBank mRNA | AF135185.1 | - | 5 - 6 | 517 | 75 | |
| AK144846.1 | - | 5 - 6 | 510 | 75 | ||
| AK161562.1 | - | 5 - 6 | 489 | 75 | ||
| BC057211.1 | - | 5 - 6 | 473 | 75 | ||
| BC064737.1 | - | 5 - 6 | 488 | 30112690 | 75 | |
| BC092526.1 | - | 5 - 6 | 537 | 6593321 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk11
Genomic Map