Assay Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011161.5 | NP_035291.4 | 8 - 9 | 768 | 67 | |
| GenBank mRNA | AF135185.1 | - | 8 - 9 | 748 | 67 | |
| AK161562.1 | - | 8 - 9 | 720 | 67 | ||
| BC057211.1 | - | 8 - 9 | 704 | 67 | ||
| BC064737.1 | - | 8 - 9 | 719 | 30112690 | 67 | |
| BC092526.1 | - | 8 - 9 | 768 | 6593321 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk11
Genomic Map