Assay Details
| Gene Symbol: Mcidas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037914.3 | NP_001033003.1 | 4 - 5 | 377 | 125 | |
| GenBank mRNA | AK134107.1 | - | 4 - 5 | 378 | 125 | |
| BC150962.1 | - | 4 - 5 | 400 | 9087873 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcidas
Genomic Map