Assay Details
Gene Symbol: Mcidas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037914.3 | NP_001033003.1 | 5 - 6 | 596 | 94 | |
GenBank mRNA | AK134107.1 | - | 5 - 6 | 597 | 94 | |
BC150962.1 | - | 5 - 6 | 619 | 9087873 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338037 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338041 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338082 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338044 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338046 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338080 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338089 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338121 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338120 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338038 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338085 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338088 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338084 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338036 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338119 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338039 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338122 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338118 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338083 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338048 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338086 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338053 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338040 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338033 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338045 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338052 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338081 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338047 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338087 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338034 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077143 | AK134107.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077143 | NM_001037914.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659775 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 252852 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561764 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535134 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647109 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109482 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077143 | AK134107.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077143 | NM_001037914.3 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109482 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109482 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338088 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338048 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338122 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338047 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338037 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338121 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338118 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338041 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338044 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338080 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338053 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338082 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338086 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338036 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338089 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338083 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338045 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338081 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338046 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338040 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338120 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338087 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338034 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338033 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338119 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338084 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338052 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338039 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338038 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29338085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338161 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11063972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10636469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338065 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10821309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338068 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10732017 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10845575 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338070 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10492251 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10942496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10942508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338061 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10746109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10779406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10845576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10760385 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10942489 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338076 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10636464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338078 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10942493 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338080 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10942500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10821307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10760386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29338072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10942510 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10407257 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407256 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407258 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407260 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407261 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407259 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10407255 | - | - |
Affymetrix Mouse Clariom S Array | PSR1300008125.mm.1 | NM_001037914.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008125.mm.1 | NM_001037914.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008125.mm.1 | TC1300001275.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300008127.mm.1 | NM_001037914.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008127.mm.1 | NM_001037914.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008127.mm.1 | TC1300001275.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300008128.mm.1 | NM_001037914.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008128.mm.1 | NM_001037914.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008128.mm.1 | TC1300001275.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300008126.mm.1 | NM_001037914.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008126.mm.1 | NM_001037914.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300008126.mm.1 | TC1300001275.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 728944 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1222308 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2995948 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2995948 | - | - |
