Assay Details
| Gene Symbol: Grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019518.3 | NP_062391.3 | 2 - 3 | 392 | 73 | |
| GenBank mRNA | AB041603.1 | - | 2 - 3 | 390 | 73 | |
| AF236099.1 | - | 2 - 3 | 355 | 73 | ||
| BC046307.1 | - | 2 - 3 | 360 | 6306542 | 73 | |
| BY730445.1 | - | 2 - 3 | 411 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grasp
Genomic Map