Assay Details
| Gene Symbol: 3830408C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015471.1 | 1 - 2 | 1414 | 67 | ||
| GenBank mRNA | AK014423.1 | - | 1 - 2 | 1414 | 67 | |
| AK043130.1 | - | 1 - 2 | 942 | 67 | ||
| AK138173.1 | - | 1 - 2 | 1432 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 2A | Kif2a |
C530030B14Rik, Kif2, Kns2, M-kinesin |
-342.0 | Anti-sense | Chr. 13 - 106960585 - 107022228 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 31452 |
Gm31452 | 37.0kb | Sense | Chr. 13 - 107063405 - 107148551 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

3830408C21Rik
Genomic Map