Assay Details
| Gene Symbol: Ampd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033303.2 | NP_001028475.2 | 12 - 13 | 1759 | 106 | |
| GenBank mRNA | AK132524.1 | - | 12 - 13 | 1750 | 106 | |
| AK137549.1 | - | 12 - 13 | 1751 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ampd1
Genomic Map