Assay Details
| Gene Symbol: Ampd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033303.2 | NP_001028475.2 | 6 - 7 | 844 | 83 | |
| GenBank mRNA | AK132524.1 | - | 6 - 7 | 835 | 83 | |
| AK137549.1 | - | 6 - 7 | 836 | 83 | ||
More Information
Set Membership: |
|

Ampd1
Genomic Map