Assay Details
| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053170.3 | NP_444400.2 | 18 - 19 | 3745 | 123 | |
| GenBank mRNA | AF220138.1 | - | 3 - 4 | 322 | 123 | |
| AY458590.1 | - | 18 - 19 | 3751 | 123 | ||
| BC043471.1 | - | 5 - 6 | 740 | 202 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim33
Genomic Map