Assay Details
| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079830.2 | NP_001073299.1 | 4 - 5 | 1549 | 100 | |
| NM_053170.3 | NP_444400.2 | 4 - 5 | 1549 | 100 | ||
| GenBank mRNA | AK129293.1 | - | 4 - 5 | 808 | 100 | |
| AY458590.1 | - | 4 - 5 | 1555 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim33
Genomic Map