Assay Details
| Gene Symbol: Lrig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025067.1 | NP_001020238.1 | 5 - 6 | 813 | 99 | |
| GenBank mRNA | AK036116.1 | - | 5 - 6 | 814 | 99 | |
| AK155089.1 | - | 5 - 6 | 876 | 99 | ||
| AK158022.1 | - | 6 - 7 | 830 | 99 | ||
| BC094228.1 | - | 5 - 6 | 750 | 30362610 | 99 | |
| BC138422.1 | - | 5 - 6 | 724 | 8861442 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrig2
Genomic Map