Assay Details
| Gene Symbol: Lrig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025067.1 | NP_001020238.1 | 7 - 8 | 1106 | 66 | |
| GenBank mRNA | AK036116.1 | - | 7 - 8 | 1108 | 66 | |
| AK155089.1 | - | 7 - 8 | 1169 | 66 | ||
| AK158022.1 | - | 8 - 9 | 1123 | 66 | ||
| BC094228.1 | - | 7 - 8 | 1043 | 30362610 | 66 | |
| BC138422.1 | - | 7 - 8 | 1017 | 8861442 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrig2
Genomic Map