Assay Details
| Gene Symbol: Hipk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301304.1 | NP_001288233.1 | 14 - 15 | 3182 | 60 | |
| NM_001301306.1 | NP_001288235.1 | 13 - 14 | 3157 | 60 | ||
| NM_010432.2 | NP_034562.2 | 14 - 15 | 3292 | 60 | ||
| NR_125731.1 | 15 - 16 | 3307 | 60 | |||
| GenBank mRNA | AF071070.1 | - | 14 - 15 | 3182 | 60 | |
| AF071071.1 | - | 15 - 16 | 3307 | 60 | ||
| AF077658.1 | - | 14 - 15 | 3193 | 60 | ||
| AF170303.1 | - | 13 - 14 | 2957 | 60 | ||
| AF170304.1 | - | 14 - 15 | 3092 | 60 | ||
| AK122333.1 | - | 7 - 8 | 1189 | 60 | ||
| BC145697.1 | - | 14 - 15 | 3045 | 40131000 | 60 | |
| BC145699.1 | - | 14 - 15 | 3045 | 40131002 | 60 | |
| DQ275150.1 | - | 3 - 4 | 480 | 60 | ||
| DQ275151.1 | - | 3 - 4 | 480 | 60 | ||
| DQ275152.1 | - | 3 - 4 | 476 | 60 | ||
| DQ275153.1 | - | 3 - 4 | 480 | 60 | ||
| DQ275154.1 | - | 3 - 4 | 480 | 60 | ||
| DQ275155.1 | - | 3 - 4 | 477 | 60 | ||
| DQ279087.1 | - | 8 - 9 | 1378 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hipk1
Genomic Map