Assay Details
| Gene Symbol: D930007P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045743.1 | 3 - 4 | 687 | 63 | ||
| GenBank mRNA | AK052984.1 | - | 3 - 4 | 688 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30528 |
Gm30528 | -63.2kb | Sense | Chr. 15 - 103026805 - 103063780 on NCBI Build GRCm38 | |
| Downstream | single-strand selective monofunctional uracil DNA glycosylase | Smug1 |
1200013B09Rik, A930006H09Rik, C85220 |
9.3kb | Sense | Chr. 15 - 103153290 - 103168895 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

D930007P13Rik
Genomic Map