Assay Details
| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172958.3 | NP_766546.1 | 2 - 3 | 247 | 76 | |
| GenBank mRNA | AK030139.1 | - | 2 - 3 | 314 | 76 | |
| AK050099.1 | - | 2 - 3 | 240 | 76 | ||
| AK050236.1 | - | 2 - 3 | 238 | 76 | ||
| AK122539.1 | - | 2 - 3 | 305 | 76 | ||
| BC042573.1 | - | 2 - 3 | 236 | 76 | ||
| BC054352.1 | - | 2 - 3 | 247 | 3661020 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr12
Genomic Map