Assay Details
| Gene Symbol: Pnpla5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029427.1 | NP_083703.1 | 8 - 9 | 1254 | 113 | |
| GenBank mRNA | AK014771.1 | - | 8 - 9 | 1255 | 113 | |
| BC109360.2 | - | 8 - 9 | 1254 | 40058461 | 113 | |
| BC109361.2 | - | 8 - 9 | 1254 | 40058463 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
biological_process
metabolic process
lipid catabolic process
molecular_function
phospholipase activity
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity
lipid metabolic process
biological_process
metabolic process
lipid catabolic process
molecular_function
phospholipase activity
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154645 | NM_029427.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154645 | AK014771.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154645 | BC109360.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395048 | AK035227.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395048 | AK053714.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395048 | BC094505.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395048 | NM_178928.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159757 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164884 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101444 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154645 | NM_029427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154645 | AK014771.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154645 | BC109360.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164884 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101444 | - | - |
| Whole Mouse Genome G4122A | A_51_P334804 | BC109361.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334804 | NM_029427.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334804 | AK014771.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P334804 | BC109360.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164884 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30392683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12324707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12324704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12440846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12404935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12134755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12404936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11975150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12141628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12191574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11910548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12343446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12252646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12141623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12404932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12355737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12065162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12191570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12343442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12355734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11914988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11908644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12141621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12324698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12141640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11910544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30392786 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10431126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431135 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431129 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430693_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10431124 | - | - |
| Mouse Genome 430 2.0 Array | 1430693_at | - | - |
| Mouse Genome 430 2.0 Array | 1430693_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75772_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75772_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500015752.mm.1 | NM_029427.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015752.mm.1 | NM_029427.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015752.mm.1 | TC1500001983.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015754.mm.1 | NM_029427.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015754.mm.1 | NM_029427.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015754.mm.1 | TC1500001983.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430693_PM_at | - | - |
| Mouse Expression Set 430 | 1430693_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430693_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430693_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1770609 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2850595 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2850595 | - | - |

Pnpla5
Genomic Map