Assay Details
| Gene Symbol: Mks1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039684.2 | NP_001034773.2 | 7 - 8 | 772 | 59 | |
| GenBank mRNA | AK030982.1 | - | 7 - 8 | 752 | 59 | |
| AK138267.1 | - | 6 - 7 | 742 | 59 | ||
| AK170482.1 | - | 7 - 8 | 772 | 59 | ||
| BC150995.1 | - | 7 - 8 | 840 | 9087908 | 59 | |
| BC158122.1 | - | 7 - 8 | 932 | 9088195 | 59 | |
| DQ177342.1 | - | 7 - 8 | 772 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mks1
Genomic Map