Assay Details
| Gene Symbol: Mks1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039684.2 | NP_001034773.2 | 8 - 9 | 77 | ||
| GenBank mRNA | AK030982.1 | - | 8 - 9 | 77 | ||
| AK170482.1 | - | 8 - 9 | 77 | |||
| AK190930.1 | - | 1 - 2 | 77 | |||
| BC150995.1 | - | 8 - 9 | 959 | 9087908 | 77 | |
| BC158122.1 | - | 8 - 9 | 1051 | 9088195 | 77 | |
| DQ177342.1 | - | 8 - 9 | 77 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
determination of left/right symmetry
regulation of smoothened signaling pathway
epithelial structure maintenance
cell projection organization
nonmotile primary cilium assembly
cilium assembly
embryonic digit morphogenesis
embryonic skeletal system development
branching morphogenesis of an epithelial tube
inner ear receptor stereocilium organization
cilium morphogenesis
head development
regulation of canonical Wnt signaling pathway
common bile duct development
regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
motile primary cilium assembly
embryonic brain development
regulation of Wnt signaling pathway, planar cell polarity pathway
protein binding
neural tube closure
determination of left/right symmetry
regulation of smoothened signaling pathway
epithelial structure maintenance
cell projection organization
nonmotile primary cilium assembly
cilium assembly
embryonic digit morphogenesis
embryonic skeletal system development
branching morphogenesis of an epithelial tube
inner ear receptor stereocilium organization
cilium morphogenesis
head development
regulation of canonical Wnt signaling pathway
common bile duct development
regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
motile primary cilium assembly
embryonic brain development
regulation of Wnt signaling pathway, planar cell polarity pathway
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27996585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27996587 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27996586 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990086 | DQ177342.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990086 | AK030982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990086 | NM_001039684.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56020 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122788 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990086 | DQ177342.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990086 | AK030982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990086 | NM_001039684.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27996581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07432293 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122788 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122788 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27996586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27996587 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27996585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27996723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09449127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09449111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09714812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09632778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09449112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09044588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09605243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09116120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09449115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09044578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09621185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09116117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27996581 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380197 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380191 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380200 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380202 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435835_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380189 | - | - |
| Mouse Genome 430 2.0 Array | 1435835_at | - | - |
| Mouse Genome 430 2.0 Array | 1435835_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380718_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380718_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100012759.mm.1 | NM_001039684.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012759.mm.1 | NM_001039684.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012759.mm.1 | TC1100001354.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012761.mm.1 | NM_001039684.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012761.mm.1 | NM_001039684.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012761.mm.1 | TC1100001354.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435835_PM_at | - | - |
| Mouse Expression Set 430 | 1435835_at | - | - |
| Murine Genome U74v2 Set | 129303_PM_at | - | - |
| Murine Genome U74v2 Set | 129303_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435835_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435835_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 475312 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231624 | - | - |

Mks1
Genomic Map