Assay Details
| Gene Symbol: Zfp213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033496.3 | NP_001028668.2 | 1 - 2 | 259 | 158 | |
| GenBank mRNA | AK133902.1 | - | 1 - 2 | 157 | 158 | |
| AK147992.1 | - | 1 - 2 | 163 | 158 | ||
| BC038178.1 | - | 1 - 2 | 172 | 158 | ||
| CJ184116.1 | - | 1 - 2 | 259 | 158 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147992.1, NM_001033496.3 transcripts.
Set Membership: |
|

Zfp213
Genomic Map