Assay Details
Gene Symbol: Zfp213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033496.3 | NP_001028668.2 | 2 - 3 | 775 | 73 | |
GenBank mRNA | AK133902.1 | - | 2 - 3 | 673 | 73 | |
AK147992.1 | - | 2 - 3 | 679 | 73 | ||
BC038178.1 | - | 2 - 3 | 688 | 73 | ||
BC150777.1 | - | 1 - 2 | 518 | 9087688 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
