Assay Details
Gene Symbol: Zfp213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033496.3 | NP_001028668.2 | 5 - 6 | 1123 | 121 | |
GenBank mRNA | AK147992.1 | - | 5 - 6 | 1027 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092404 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092271 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092401 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092273 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092360 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092274 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092362 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092365 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092361 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092363 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092406 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092272 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092402 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092364 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092275 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092405 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P09657 | BX528154.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P09657 | AK147992.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P09657 | AK171979.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P09657 | NM_001033496.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P09657 | AK133902.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738722 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366621 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689770 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865044 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51665 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P09657 | BX528154.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P09657 | AK147992.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P09657 | AK171979.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P09657 | NM_001033496.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P09657 | AK133902.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08736980 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08736998 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092365 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092401 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092273 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092404 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092402 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092363 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092362 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092405 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092275 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092361 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092364 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092271 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092360 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092274 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092406 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31092272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31092366 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13461513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13187799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092359 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092345 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092402 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092400 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13585756 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092360 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092358 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13585753 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13424505 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13229886 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092365 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092283 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13466958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13335989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13461508 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13351924 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092380 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092372 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092323 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13507975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092353 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092397 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092385 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13229858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092370 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13507972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13461497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092349 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092376 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13229867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13210274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13085028 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13229881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092393 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092346 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13187802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092280 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092410 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13229860 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13466968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31092366 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13351913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13498457 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10448265 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448267 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448264 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448263 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448268 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10448266 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438479_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1433705_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10448262 | - | - |
Mouse Genome 430 2.0 Array | 1433705_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1433705_at | - | - |
Mouse Genome 430 2.0 Array | 1438479_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1438479_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 449521_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 449521_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700013898.mm.1 | NM_001033496.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013898.mm.1 | NM_001033496.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013898.mm.1 | TC1700001593.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700013897.mm.1 | NM_001033496.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013897.mm.1 | NM_001033496.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700013897.mm.1 | TC1700001593.mm.1 | MTA TC |
Mouse Expression Set 430 | 1433705_at | - | - |
Mouse Expression Set 430 | 1433705_PM_at | - | - |
Mouse Expression Set 430 | 1438479_at | - | - |
Mouse Expression Set 430 | 1438479_PM_at | - | - |
Murine Genome U74v2 Set | 101369_PM_at | - | - |
Murine Genome U74v2 Set | 101369_at | - | - |
Mouse Genome 430A 2.0 Array | 1433705_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1433705_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1433705_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1433705_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438479_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438479_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 535414 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160203 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3160203 | - | - |
mouse-6 v1.1 (Illumina) | GI_85702030-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3160203 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160203 | - | - |
