Assay Details
| Gene Symbol: Rgs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282061.1 | NP_001268990.1 | 6 - 7 | 474 | 67 | |
| NM_015812.3 | NP_056627.1 | 6 - 7 | 756 | 67 | ||
| GenBank mRNA | AF061933.1 | - | 3 - 4 | 201 | 67 | |
| AK052527.1 | - | 6 - 7 | 757 | 67 | ||
| AK053087.1 | - | 6 - 7 | 475 | 67 | ||
| AK139323.1 | - | 6 - 7 | 871 | 67 | ||
| BC038616.1 | - | 4 - 5 | 301 | 67 | ||
| BC125029.1 | - | 6 - 7 | 460 | 40062355 | 67 | |
| BC125030.1 | - | 6 - 7 | 460 | 40062356 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgs6
Genomic Map