Assay Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008372.4 | NP_032398.3 | 2 - 3 | 355 | 129 | |
| GenBank mRNA | AF078906.1 | - | 2 - 3 | 221 | 129 | |
| AK040364.1 | - | 2 - 3 | 338 | 129 | ||
| AK040740.1 | - | 2 - 3 | 338 | 129 | ||
| AK041838.1 | - | 2 - 3 | 246 | 129 | ||
| AK042264.1 | - | 2 - 3 | 300 | 129 | ||
| AK154516.1 | - | 2 - 3 | 315 | 129 | ||
| BC089571.1 | - | 2 - 3 | 307 | 6758487 | 129 | |
| M29697.1 | - | 2 - 3 | 268 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il7r
Genomic Map