Assay Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008372.4 | NP_032398.3 | 6 - 7 | 937 | 151 | |
| GenBank mRNA | AF078906.1 | - | 6 - 7 | 803 | 151 | |
| AK040364.1 | - | 6 - 7 | 920 | 151 | ||
| AK042264.1 | - | 6 - 7 | 882 | 151 | ||
| AK154215.1 | - | 5 - 6 | 751 | 151 | ||
| AK154516.1 | - | 6 - 7 | 152 | |||
| BC089571.1 | - | 6 - 7 | 889 | 6758487 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il7r
Genomic Map