Assay Details
| Gene Symbol: B4galt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020579.2 | NP_065604.2 | 1 - 2 | 85 | 82 | |
| GenBank mRNA | BC004095.1 | - | 1 - 2 | 48 | 82 | |
| BC013619.1 | - | 1 - 2 | 59 | 3965561 | 82 | |
| BU703242.1 | - | 1 - 2 | 85 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC013619.1, NM_020579.2 transcripts.
Set Membership: |
|

B4galt3
Genomic Map