Assay Details
| Gene Symbol: B4galt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020579.2 | NP_065604.2 | 4 - 5 | 788 | 63 | |
| GenBank mRNA | AF142671.1 | - | 4 - 5 | 728 | 63 | |
| AK155471.1 | - | 5 - 6 | 888 | 63 | ||
| BC004095.1 | - | 4 - 5 | 547 | 63 | ||
| BC013619.1 | - | 4 - 5 | 761 | 3965561 | 63 | |
| BC021830.1 | - | 2 - 3 | 1584 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galt3
Genomic Map