Assay Details
| Gene Symbol: Rpap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028003.2 | NP_082279.1 | 1 - 2 | 75 | 64 | |
| GenBank mRNA | AK009765.1 | - | 1 - 2 | 76 | 64 | |
| BC004046.1 | - | 1 - 2 | 78 | 3496407 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK009765.1,BC004046.1,NM_028003.2 transcripts.
This assay is in the 5'UTR region of the AK009765.1,BC004046.1,NM_028003.2 transcripts.
Set Membership: |
|

Rpap3
Genomic Map