Assay Details
| Gene Symbol: Pfkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163487.1 | NP_001156959.1 | 8 - 9 | 897 | 87 | |
| NM_001163488.1 | NP_001156960.1 | 8 - 9 | 833 | 87 | ||
| NM_021514.4 | NP_067489.3 | 7 - 8 | 850 | 87 | ||
| GenBank mRNA | AF249894.1 | - | 7 - 8 | 790 | 87 | |
| AK002711.2 | - | 8 - 9 | 800 | 87 | ||
| AK049773.1 | - | 8 - 9 | 805 | 87 | ||
| BC005526.1 | - | 7 - 8 | 814 | 3495173 | 87 | |
| BC115774.1 | - | 5 - 6 | 605 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkm
Genomic Map