Assay Details
| Gene Symbol: Endou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008902.3 | NP_032928.1 | 1 - 2 | 311 | 95 | |
| GenBank mRNA | M95545.1 | - | 1 - 2 | 311 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Endou| Gene Symbol: Endou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008902.3 | NP_032928.1 | 1 - 2 | 311 | 95 | |
| GenBank mRNA | M95545.1 | - | 1 - 2 | 311 | 95 | |
Set Membership: |
|