Assay Details
| Gene Symbol: Vdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009504.4 | NP_033530.2 | 1 - 2 | 64 | 60 | |
| GenBank mRNA | AK030339.1 | - | 1 - 2 | 65 | 60 | |
| AK154647.1 | - | 1 - 2 | 39 | 60 | ||
| AK156789.1 | - | 1 - 2 | 80 | 60 | ||
| D31969.1 | - | 1 - 2 | 32 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030339.1, AK154647.1, AK156789.1, D31969.1, NM_009504.4 transcripts.
Set Membership: |
|

Vdr
Genomic Map