Assay Details
| Gene Symbol: Vdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009504.4 | NP_033530.2 | 2 - 3 | 132 | 92 | |
| GenBank mRNA | AK030339.1 | - | 2 - 3 | 133 | 92 | |
| AK154647.1 | - | 2 - 3 | 107 | 92 | ||
| AK156789.1 | - | 2 - 3 | 148 | 92 | ||
| BC006716.1 | - | 2 - 3 | 98 | 3710866 | 92 | |
| D31969.1 | - | 2 - 3 | 100 | 92 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK030339.1,AK154647.1,AK156789.1,BC006716.1,D31969.1,NM_009504.4 transcripts.
Set Membership: |
|

Vdr
Genomic Map