Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100011585.mm.1 |
NM_178680.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011585.mm.1 |
NM_178680.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011585.mm.1 |
TC1100001235.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100011588.mm.1 |
NM_178680.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011588.mm.1 |
NM_178680.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011588.mm.1 |
TC1100001235.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100011586.mm.1 |
NM_178680.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011586.mm.1 |
NM_178680.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011586.mm.1 |
TC1100001235.mm.1 |
MTA TC |