Assay Details
| Gene Symbol: Unc45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178680.4 | NP_848795.3 | 1 - 2 | 110 | 80 | |
| GenBank mRNA | AK084418.1 | - | 1 - 2 | 78 | 80 | |
| AK168968.1 | - | 1 - 2 | 78 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084418.1, NM_178680.4 transcripts.
Set Membership: |
|

Unc45b
Genomic Map