Assay Details
| Gene Symbol: Higd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002900.1 | NP_001002900.1 | 1 - 2 | 45 | 65 | |
| GenBank mRNA | AB099517.1 | - | 1 - 2 | 45 | 65 | |
| AB253942.1 | - | 1 - 2 | 85 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB099517.1, AB253942.1, NM_001002900.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046666 | AB099516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046666 | AK144032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046666 | NM_001024672.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046666 | AB110950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046666 | AB253942.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109190 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136975 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144641 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046666 | AB099516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046666 | AK144032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046666 | NM_001024672.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046666 | AB110950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046666 | AB253942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P719962 | AB099516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P719962 | AK144032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P719962 | NM_001024672.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P719962 | AB110950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P719962 | AB253942.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11838771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11923664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12101050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11983544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30485376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11863852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12466785 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426903 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426901 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426898 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500008264.mm.1 | NM_001002900.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008264.mm.1 | NM_001002900.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008264.mm.1 | TC1500001044.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456510_PM_x_at | - | - |
| Mouse Expression Set 430 | 1434150_a_at | - | - |
| Mouse Expression Set 430 | 1434150_PM_a_at | - | - |
| Mouse Expression Set 430 | 1421184_a_at | - | - |
| Mouse Expression Set 430 | 1456510_x_at | - | - |
| Mouse Expression Set 430 | 1421184_PM_a_at | - | - |
| Mouse Expression Set 430 | 1447356_PM_at | - | - |
| Mouse Expression Set 430 | 1447356_at | - | - |
| Murine Genome U74v2 Set | 131101_PM_at | - | - |
| Murine Genome U74v2 Set | 131101_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421184_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434150_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434150_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456510_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456510_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421184_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215033 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2819924 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2819924 | - | - |

Higd1c
Genomic Map