Assay Details
| Gene Symbol: Scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF330052.1 | - | 7 - 8 | 686 | 140 | |
| AK186804.1 | - | 4 - 5 | 244 | 140 | ||
| AK188859.1 | - | 4 - 5 | 241 | 140 | ||
| BC004847.1 | - | 7 - 8 | 720 | 2650587 | 140 | |
| BC021399.1 | - | 7 - 8 | 685 | 4193674 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scpep1
Genomic Map