Assay Details
| Gene Symbol: Scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC004847.1 | - | 9 - 10 | 937 | 2650587 | 90 |
| BC021399.1 | - | 9 - 10 | 902 | 4193674 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scpep1
Genomic Map