Assay Details
| Gene Symbol: Slc35f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028787.4 | NP_083063.2 | 10 - 11 | 1219 | 71 | |
| GenBank mRNA | AK004892.2 | - | 10 - 11 | 1200 | 71 | |
| AK076339.1 | - | 10 - 11 | 1213 | 71 | ||
| AK145848.1 | - | 10 - 11 | 1220 | 71 | ||
| AK169012.1 | - | 10 - 11 | 1081 | 71 | ||
| AK171951.1 | - | 10 - 11 | 1193 | 71 | ||
| BC026858.1 | - | 10 - 11 | 1078 | 4014064 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35f5
Genomic Map